Anti-Connexin 43 Rabbit Polyclonal Antibody [clone: S369]
BWRLBS1073
- Antibody type:Primary
- Antigen symbol:Connexin 43
- Clonality:Polyclonal
- Clone:S369
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 340-390 of Human Connexin 43.
Connexin 43 (S369) pAb detects endogenous levels of Connexin 43 protein.
Type: Primary
Antigen: Connexin 43
Clonality: Polyclonal
Clone: S369
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat