B2m (Rat) ELISA kit
Catalog # ABNOKA2021
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Rat
- Description:B2m (Rat) ELISA Kit, 96 assays
- Tests per kit:96 assays
Specifications
About this item
B2m (Rat) ELISA Kit is an enzyme immunoassay used for the quantitative determination of Beta 2-Microglobulin (B2m) in rat biological fluids.
- Calibration range: 0,156 to 5 ng/ml
- Sample volume: 100 µl
Delivery: Kit contains Anti-Rat B2M ELISA 96-well microplate (8×12), calibrator (1 vial), running buffer (5×50 ml), washing solution concentrate (20×50 ml), enzyme-antibody conjugate (100×150 µl), chromogen-substrate solution (12 ml) and stop solution (12 ml).