Agarose multipurpose, white powder for molecular biology
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:9012-36-6
:
MDL Number:MFCD00081294
Melting Pt:89 °C ± 1.5 °C (1.5% gel)
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Gelling Temperature (1.5% gel):36 ± 1.5°C
- Melting Temperature (1.5% gel):89 ± 1.5°C
- Gel Strength (1.5% gel):> 3200 g/cm2
- Electroendosmosis:<0.12%
- Sulfate:<0.07%
- DNase/Rnase:None Detected
- DNA Blotting:Complete Transfer
- Inhibition of Ligation and Restriction Enzymes:None Detected
Specification Test Results
Frequently Bought Together