Guanidinium thiocyanate ≥98%
Catalog # ICNA04820991
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:118.16 g/mol
:GHS05
:GHS07
Storage Temperature:Ambient
CAS Number:593-84-0
Formula:CH₅N₃·CHNS
MDL Number:MFCD00013027