Sodio alginato (da Macrocystis pyrifera (kelp)), off-white powder, low viscosity
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:9005-38-3
:
MDL Number:MFCD00081310
Storage Temperature:Ambiente
Specification Test Results
- Identity Test:Passes
- Source:Macrocystis pyrifera (Kelp)
- Viscosity (1% aq soln):≤300 cps
Specification Test Results
Frequently Bought Together