Crotalus adamanteus Phosphodiesterase I, MP Biomedicals
Catalog # ICNA0210097880
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:100 AU
- CAS No.:9025-82-5
- Species:Crotalus adamanteus
- Storage conditions:−20 °C
- Enzyme name:Phosphodiesterase I
- Molecular weight:115,000
- pH:Optimum pH=9.8 to 10.4.5(Lit.)
Specifications
About this item
Lyophilised in vials containing approximately 5 mg dry wt. and at least 100 units/vial (20 to 40 units/mg dry wt.).
Unit definition: One unit hydrolyses 1 µmole of p-nitrophenyl thymidine-5-phosphate per minute at pH 8,9 and 25 °C
pH: optimum pH=9,8 to 10,4.5 (Lit.)