Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Italy
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Aluminium oxide
Aluminium oxide, Ecochrom™ Alumina B Activity: Super I
Aluminium oxide, Ecochrom™ Alumina B Activity: Super I
ICNA0204571
:
MP Biomedicals
:
Aluminium oxide, Ecochrom™ Alumina B Activity: Super I
ICNA0204571
:
MP Biomedicals
:
Pk:
1 kg
Seq:
4024
:
:
101.96 g/mol
:
3.94 g/cm³ (20 °C)
:
Ambient
:
1344-28-1
:
11092-32-3
:
Al₂O₃
:
MFCD00003424