Silicon dioxide, Silitech 18-32 µm
Catalog #
Product Details & Documents
- Mean Pore Size: 60 A
- Particle Size: 18-32 µm
pH: 7.0 ± 0.5
Synonyms: Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
- CAS Number:7631-86-9
- Formula:SiO₂
- MDL Number:MFCD00011232
- Molecular Weight:60.08 g/mol
- Density:∼2 g/cm³ (20 °C) (lit.)
- Storage Temperature:Ambient



