Flame retardant overalls, MICROGARD® 1500 PLUS FR, model 111
Supplier: Ansell
Certificates
About this item
These overalls are made from a highly breathable, flame retardant and anti-static SMMS polypropylene non woven fabric designed for protection from particulates and light, non flammable liquid spray or splash.
- Three-piece hood
- Two-way front zipper with resealable storm flap; finger loops
- Elasticated wrists, waist and ankles
- Red stitching for ease of identification
- Stitched seams combine strength with particle barrier
- Flame retardant and anti-static SMMS provides a good barrier to particulates and low hazard liquid sprays or splashes
- Air and water vapour permeable ('breathable') to help reduce the risk of heat stress
Intended to be worn over heat and flame protective clothing, MICROGARD® 1500 PLUS FR offers wearers protection from particulates (Type 5) and non flammable, non pressurised liquid sprays or splashes (Type 6).*
Typical applications include: Petrochemical industry, industrial cleaning, utilities, general maintenance.
Certifications: Type 5/6; EN 1073-2; EN ISO 14116 (index 1/0/0), EN 1149-5; Cat. III
Caution: Must be worn over thermal protective garments (which achieve EN ISO 14116 Index 2 or above), such as NOMEX® or PYROVATEX®, and never be worn next to the skin.
Specifications
- Catalog No:
- ANSEWR17S-00111-07
- ANSEWR17S-00111-06
- ANSEWR17S-00111-05
- ANSEWR17S-00111-04
- ANSEWR17S-00111-03
- Model:
- 111
- 111
- 111
- 111
- 111
- Anti-static:
- +
- +
- +
- +
- +
- Garment application:
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial
- Chemical protection, Industrial