ESD safety ankle boots, lace-up, E-Sport, JALAS® 1625
Supplier: EJENDALS
Certificates
About this item
Anti-static, lightweight, ESD safety ankle boots made from PU coated split leather with EVA, polyester-based conductive thread insole, polyester/polyamide lining plus a PU outsole.
- Orthopaedic insoles possible
- Ergothan shock absorption zone
- Toe protection cap made from aluminium
- Flexible, puncture resistant insole made from plasma-treated composite (PTC)
Mainly used for outdoors, indoors and environments with risk for outsole penetration.
Colour: Black/grey/red
Certifications: EN 61340-5-1 (ESD); BGR191; EN ISO 20345:2011; S3, SRC, CI
Frequently Bought Together