ESD safety ankle boots, lace-up, Jupiter, Jalas® 1828
Supplier: EJENDALS
Certificates
About this item
Wide fit, anti-static ESD safety ankle boots made from PU coated leather with EVA, polyester-based conductive thread insole, polyester/polyamide lining plus a heat and oil resistant outsole.
- Double shock-absorption zones in Poron® XRD®
- ProNose toe reinforcement
- Puncture resistant steel midsole
- Toe protection cap made from aluminium (200 J)
Mainly used for outdoors, environments with risk for outsole penetration and harsh environments.
Colour: Black/grey/yellow
Certifications: EN 61340-5-1 (ESD); EN ISO 20345:2011; S3, SRC, CI, HRO
Frequently Bought Together