Order Entry
Italy
ContactUsLinkComponent
Chromeazurol S (Mordant blue 29) indicator for metal titration, Fluka™
Chromeazurol S (Mordant blue 29) indicator for metal titration, Fluka™
  72687-25G
 :  
Chromeazurol S (Mordant blue 29) indicator for metal titration, Fluka™
  72687-25G
 :  
  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308
  • Identity (IR):
    complying
  • Extinction maximum at:
    424 - 436 nm
  • Molar absorbancy index:
    Min. 6000
 :
 :605.28 g/mol
 :Ambient
 :216-787-0
 :1667-99-8
 :C₂₃H₁₆Cl₂O₉S
 :MFCD00001615