Cut resistant gloves, HyFlex® 11-727
Supplier: Ansell
Certificates
About this item
These knitted seamless liners with INTERCEPT™ technology yarn and PU coated palm combine excellent comfort and mobility with optimal cut protection.
- Best-in-class cut protection and increased comfort and dexterity
- Palm coating design improves worker grip and extends glove life in abrasive conditions
- Uncoated back allows the hand to breath
- Grey coating hides dirt in environments with dirt exposure
INTERCEPT™ is a specialised knitting technology which blends engineered, synthetic and natural fibres into high performance yarns. This provides high-cut protection with exceptional comfort and dexterity.
Ideal for assembling white goods, general maintenance, glass manufacturing, medical device manufacturing, plastics injection and moulding, and pre-assembling. Also suitable for handling sharp-edged objects and for cutting small dry or lightly oiled parts.
Certifications: EN 388:2016 (4342B); Cat. II
Specifications
- Catalog No:
- ANSE11-727/11
- ANSE11-727/10
- ANSE11-727/7
- ANSE11-727/5
- ANSE11-727/6
- ANSE11-727/9
- ANSE11-727/8
- Model:
- 11-727
- 11-727
- 11-727
- 11-727
- 11-727
- 11-727
- 11-727
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Finish:
- Palm dipped
- Palm dipped
- Palm dipped
- Palm dipped
- Palm dipped
- Palm dipped
- Palm dipped
- Gauge:
- 15
- 15
- 15
- 15
- 15
- 15
- 15
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)
- EN 388:2016 (4342B)