Order Entry
Italy
ContactUsLinkComponent
Anti-KRT76 Monoclonal Antibody [clone: KRTH/1576R]
Anti-KRT76 Monoclonal Antibody [clone: KRTH/1576R]
  ABNORAB00380
 :  Abnova
Anti-KRT76 Monoclonal Antibody [clone: KRTH/1576R]
  ABNORAB00380
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Keratin 76
  • Antigen symbol:
    KRT76
  • Clonality:
    Monoclonal
  • Clone:
    KRTH/1576R
  • Conjugation:
    Unconjugated
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    51350
  • Storage buffer:
    In 10 mM PBS.
  • Storage temperature:
    Store at -20 to -80° C.
  • Immunogen:
    Recombinant KRT76.
  • Purification:
    Protein A/G purification
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recombinant rabbit monoclonal antibody raised against human KRT76.

Type: Primary
Antigen: KRT76
Clonality: Monoclonal
Clone: KRTH/1576R
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG, kappa
Reactivity: Human