Order Entry
Italy
ContactUsLinkComponent
Anti-ITGB3 Mouse Monoclonal Antibody [clone: Y2/51]
Anti-ITGB3 Mouse Monoclonal Antibody [clone: Y2/51]
Catalog # ABNOMAB14568
Supplier:  Abnova
Anti-ITGB3 Mouse Monoclonal Antibody [clone: Y2/51]
Catalog # ABNOMAB14568
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Phytohemagglutinin (PHA)-stimulated peripheral blood cells
  • Antigen symbol:
    ITGB3
  • Clonality:
    Monoclonal
  • Clone:
    Y2/51
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    3690
  • Storage buffer:
    In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
  • Storage temperature:
    Store at 4° C.
  • Immunogen:
    Phytohemagglutinin (PHA)-stimulated peripheral blood cells.
  • Purification:
    Protein A/G purification
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal antibody raised against native human ITGB3.

Type: Primary
Antigen: ITGB3
Clonality: Monoclonal
Clone: Y2/51
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human