Anti-Human KAL cells Mouse Monoclonal Antibody [clone: KBA.62]
Catalog # ABNOMAB14429
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Human KAL cells derived from lymph node metastasis of malignant melanoma
- Clonality:Monoclonal
- Clone:KBA.62
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Storage buffer:In 10 mM PBS (0.05% BSA and 0.05% azide).
- Storage temperature:Store at 4° C.
- Immunogen:Human KAL cells derived from lymph node metastasis of malignant melanoma.
- Purification:Protein A/G purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against human melanoma.
Type: Primary
Antigen:
Clonality: Monoclonal
Clone: KBA.62
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human