GraceResolv™ high-resolution flash cartridges
Supplier: Alltech
Certificates
About this item
GraceResolv™ cartridges are packed with a grade of Davisil® silica designed to maximise performance of today's flash instruments.
- High resolution
- Increased loading capacity
- Reproducible performance
- Compatible with all flash instruments
Frequently Bought Together