Order Entry
Italy
Orders LinkContactUsLinkComponent
Selfpacker® Preparative HPLC Columns
Selfpacker® Preparative HPLC Columns
Selfpacker® Preparative HPLC Columns
Certificates
Selfpacker® Preparative HPLC Columns
Selfpacker® Preparative HPLC Columns
61 Options Available Below
Product Details & Documents
This column-packing technology is based on a combination of vacuum suction with subsequent column bed compression that stabilises the packing. A common water-jet pump is used to generate the vacuum. The sorbent slurry is sucked through a packing reservoir into the column by applying low pressure at the bottom end of the column.
  • Allows precise adjustment of the desired packing volume or sorbent bed height
  • Facilitates the determination of the column geometry
  • Makes direct scale-up from analytical to preparative scale easy
Packing stand NW25
Min./max. packing height: 185/220 mm
Max. working pressure: 200 bar
Packing stand NW50
Min./max. packing height: 185/220 mm
Max. working pressure: 100 bar
Packing stand NW100
Min./max. packing height: 295/350 mm
Max. working pressure: 100 bar
Max. working pressure: 200 bar
Product Family Options
Packing stand NW25
Product Information
  • Ø int.
    Length
    Availability
    Price
  • 556-0001
    Frit holder for compression piston (included in 556-0017)
    Frit holder for compression piston (included in 556-0017)
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 457,00 €
    457,00 €
    1 * 1 pezzi
       457,00 €
  • 556-0002
    Sealing ring for compression piston (included in 556-0017)
    Sealing ring for compression piston (included in 556-0017)
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 165,00 €
    165,00 €
    1 * 1 pezzi
       165,00 €
  • 556-0003
    Gasket set (top flange) for packing stand NW25 (included in 556-0017)
    Gasket set (top flange) for packing stand NW25 (included in 556-0017)
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 163,00 €
    163,00 €
    1 * 1 pezzi
       163,00 €
  • 556-0073
    PEEK connection set for NW25 and NW50
    PEEK connection set for NW25 and NW50
    Log in for availability
    Log in for availability
    1 * 5 pezzi
    1 * 5 pezzi - 167,00 €
    167,00 €
    1 * 5 pezzi
       167,00 €
  • 556-0021
    Hydraulic stand
    Hydraulic stand
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 6 690,00 €
    6 690,00 €
    1 * 1 pezzi
       6 690,00 €
  • 556-0029
    Tripod for NW25 and NW50 columns
    Tripod for NW25 and NW50 columns
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 1 160,00 €
    1 160,00 €
    1 * 1 pezzi
       1 160,00 €
  • 556-0032
    Packing tube 25 mm I-Ø
    Packing tube 25 mm I-Ø
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 296,00 €
    296,00 €
    1 * 1 pezzi
       296,00 €
  • 556-0019
    Complete column tube without hydraulics
    Complete column tube without hydraulics
    25,00 mm
    155 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 4 270,00 €
    4 270,00 €
    1 * 1 pezzi
       4 270,00 €
  • 556-0035
    Column tube
    Column tube
    25,00 mm
    165 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 639,00 €
    639,00 €
    1 * 1 pezzi
       639,00 €
  • 556-0036
    Column tube with temperature control jacket
    Column tube with temperature control jacket
    25,00 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 2 020,00 €
    2 020,00 €
    1 * 1 pezzi
       2 020,00 €
  • 556-0038
    Column with temperature control jacket
    Column with temperature control jacket
    25,00 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 5 880,00 €
    5 880,00 €
    1 * 1 pezzi
       5 880,00 €
  • 556-0017
    Packing stand NW25 complete with hydraulics stand
    Packing stand NW25 complete with hydraulics stand
    25,00 mm
    260 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 10 530,00 €
    10 530,00 €
    1 * 1 pezzi
       10 530,00 €
  • 556-0018
    Complete column tube without hydraulics
    Complete column tube without hydraulics
    25,00 mm
    260 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 4 730,00 €
    4 730,00 €
    1 * 1 pezzi
       4 730,00 €
  • 556-0024
    Complete column tube with temperature control jacket without hydraulics
    Complete column tube with temperature control jacket without hydraulics
    25,00 mm
    260 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 6 230,00 €
    6 230,00 €
    1 * 1 pezzi
       6 230,00 €
  • 556-0034
    Column tube with temperature control jacket
    Column tube with temperature control jacket
    25,00 mm
    260 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 2 040,00 €
    2 040,00 €
    1 * 1 pezzi
       2 040,00 €
  • 556-0030
    Packing stand NW25 complete with hydraulics stand
    Packing stand NW25 complete with hydraulics stand
    25,00 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 8 830,00 €
    8 830,00 €
    1 * 1 pezzi
       8 830,00 €
  • 556-0033
    Column tube with temperature control jacket
    Column tube with temperature control jacket
    25,00 mm
    340 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 2 250,00 €
    2 250,00 €
    1 * 1 pezzi
       2 250,00 €
  • Column
    25,00 mm
    400 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 5 580,00 €
    5 580,00 €
    1 * 1 pezzi
       5 580,00 €
  • 556-0039
    Column tube with temperature control jacket
    Column tube with temperature control jacket
    25,00 mm
    400 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 7 430,00 €
    7 430,00 €
    1 * 1 pezzi
       7 430,00 €
  • 556-0031
    Packing stand NW25 complete with hydraulics stand
    Packing stand NW25 complete with hydraulics stand
    25,00 mm
    400 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 9 640,00 €
    9 640,00 €
    1 * 1 pezzi
       9 640,00 €
Packing stand NW50
Product Information
  • Ø int.
    Length
    Availability
    Price
  • 556-0000
    Fitting set for packing stand NW50 (included in 556-0006 and 556-0011)
    Fitting set for packing stand NW50 (included in 556-0006 and 556-0011)
    — mm
    — mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 552,00 €
    552,00 €
    1 * 1 pezzi
       552,00 €
  • 556-0004
    Frit holder for compression piston (included in 556-0006 and 556-0011)
    Frit holder for compression piston (included in 556-0006 and 556-0011)
    — mm
    — mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 309,00 €
    309,00 €
    1 * 1 pezzi
       309,00 €
  • 556-0005
    Sealing ring, 50 mm (included in 556-0006 and 556-0011)
    Sealing ring, 50 mm (included in 556-0006 and 556-0011)
    — mm
    — mm
    Log in for availability
    Log in for availability
    1 * 2 pezzi
    1 * 2 pezzi - 195,00 €
    195,00 €
    1 * 2 pezzi
       195,00 €
  • 556-0072
    Gasket set (top flange), (included in 556-0006 and 556-0011)
    Gasket set (top flange), (included in 556-0006 and 556-0011)
    — mm
    — mm
    Log in for availability
    Log in for availability
    1 * 2 pezzi
    1 * 2 pezzi - 355,00 €
    355,00 €
    1 * 2 pezzi
       355,00 €
  • 556-0020
    Complete column tube top flange with screw seal without hydraulics stand
    Complete column tube top flange with screw seal without hydraulics stand
    50,00 mm
    155 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 4 320,00 €
    4 320,00 €
    1 * 1 pezzi
       4 320,00 €
  • 556-0045
    Column tube
    Column tube
    50,00 mm
    155 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 746,00 €
    746,00 €
    1 * 1 pezzi
       746,00 €
  • 556-0041
    Column tube with temperature control jacket
    Column tube with temperature control jacket
    50,00 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 2 290,00 €
    2 290,00 €
    1 * 1 pezzi
       2 290,00 €
  • 556-0058
    Column with temperature control jacket and chain closing system
    Column with temperature control jacket and chain closing system
    50,00 mm
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 7 040,00 €
    7 040,00 €
    1 * 1 pezzi
       7 040,00 €
  • 556-0006
    Complete packing stand NW50 top flange with screw seal, incl. hydraulics stand
    Complete packing stand NW50 top flange with screw seal, incl. hydraulics stand
    50,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 9 690,00 €
    9 690,00 €
    1 * 1 pezzi
       9 690,00 €
  • 556-0007
    Complete column tube top flange with screw seal without hydraulics
    Complete column tube top flange with screw seal without hydraulics
    50,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 4 440,00 €
    4 440,00 €
    1 * 1 pezzi
       4 440,00 €
  • 556-0011
    Complete packing stand NW50 with top flange chain bolted, incl. hydraulics stand
    Complete packing stand NW50 with top flange chain bolted, incl. hydraulics stand
    50,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 10 890,00 €
    10 890,00 €
    1 * 1 pezzi
       10 890,00 €
  • 556-0012
    Complete column tube with top flange chain bolted without hydraulics
    Complete column tube with top flange chain bolted without hydraulics
    50,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 5 710,00 €
    5 710,00 €
    1 * 1 pezzi
       5 710,00 €
  • 556-0022
    Complete column tube with temperature control jacket and top flange with screw seal without hydraulics stand
    Complete column tube with temperature control jacket and top flange with screw seal without hydraulics stand
    50,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 7 580,00 €
    7 580,00 €
    1 * 1 pezzi
       7 580,00 €
  • 556-0023
    Complete column tube with temperature control jacket and top flange chain bolted without hydraulics
    Complete column tube with temperature control jacket and top flange chain bolted without hydraulics
    — mm
    — mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi
  • 556-0040
    Column tube
    Column tube
    50,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 938,00 €
    938,00 €
    1 * 1 pezzi
       938,00 €
  • 556-0042
    Column tube with temperature control jacket
    Column tube with temperature control jacket
    50,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 2 370,00 €
    2 370,00 €
    1 * 1 pezzi
       2 370,00 €
  • 556-0047
    Column tube
    Column tube
    50,00 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 1 160,00 €
    1 160,00 €
    1 * 1 pezzi
       1 160,00 €
  • Column
    50,00 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 14 190,00 €
    14 190,00 €
    1 * 1 pezzi
       14 190,00 €
  • 556-0046
    Column tube
    Column tube
    50,00 mm
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 1 200,00 €
    1 200,00 €
    1 * 1 pezzi
       1 200,00 €
  • 556-0049
    Column tube with temperature control jacket
    Column tube with temperature control jacket
    50,00 mm
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 2 770,00 €
    2 770,00 €
    1 * 1 pezzi
       2 770,00 €
  • 556-0052
    Column with temperature control jacket and chain closing system
    Column with temperature control jacket and chain closing system
    50,00 mm
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 8 450,00 €
    8 450,00 €
    1 * 1 pezzi
       8 450,00 €
  • 556-0053
    Column with chain closing system
    Column with chain closing system
    50,00 mm
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 7 000,00 €
    7 000,00 €
    1 * 1 pezzi
       7 000,00 €
  • Column
    50,00 mm
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 5 220,00 €
    5 220,00 €
    1 * 1 pezzi
       5 220,00 €
  • 556-0055
    Column with temperature control jacket
    Column with temperature control jacket
    50,00 mm
    350 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 7 500,00 €
    7 500,00 €
    1 * 1 pezzi
       7 500,00 €
  • 556-0044
    Column tube
    Column tube
    50,00 mm
    400 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 1 850,00 €
    1 850,00 €
    1 * 1 pezzi
       1 850,00 €
  • 556-0048
    Column tube with temperature control jacket
    Column tube with temperature control jacket
    50,00 mm
    400 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 2 930,00 €
    2 930,00 €
    1 * 1 pezzi
       2 930,00 €
  • Column
    50,00 mm
    400 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 5 860,00 €
    5 860,00 €
    1 * 1 pezzi
       5 860,00 €
  • 556-0051
    Column tube
    Column tube
    50,00 mm
    500 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 2 510,00 €
    2 510,00 €
    1 * 1 pezzi
       2 510,00 €
  • 556-0057
    Column, 500 bar
    Column, 500 bar
    50,00 mm
    500 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 6 430,00 €
    6 430,00 €
    1 * 1 pezzi
       6 430,00 €
  • 556-0043
    Column tube
    Column tube
    50,00 mm
    650 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 2 650,00 €
    2 650,00 €
    1 * 1 pezzi
       2 650,00 €
  • 556-0050
    Column tube
    Column tube
    50,00 mm
    1000 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 4 080,00 €
    4 080,00 €
    1 * 1 pezzi
       4 080,00 €
Packing stand NW100
Product Information
  • Ø int.
    Length
    Availability
    Price
  • 556-0013
    Frit holder for packing stand NW100
    Frit holder for packing stand NW100
    — mm
    — mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 1 880,00 €
    1 880,00 €
    1 * 1 pezzi
       1 880,00 €
  • 556-0014
    Sealing ring 100 mm
    Sealing ring 100 mm
    — mm
    — mm
    Log in for availability
    Log in for availability
    1 * 2 pezzi
    1 * 2 pezzi - 427,00 €
    427,00 €
    1 * 2 pezzi
       427,00 €
  • 556-0074
    Gasket set (top flange), 1 frit, 1 PTFE ring, 1 distribution plate
    Gasket set (top flange), 1 frit, 1 PTFE ring, 1 distribution plate
    — mm
    — mm
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 700,00 €
    700,00 €
    1 * 1 SET
       700,00 €
  • 556-0016
    PTFE tubing 1,5 m × 1/4 with stainless steel jacket
    PTFE tubing 1,5 m × 1/4 with stainless steel jacket
    — mm
    — mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 603,00 €
    603,00 €
    1 * 1 pezzi
       603,00 €
  • 556-0028
    O-ring set for frit holder NW100
    O-ring set for frit holder NW100
    — mm
    — mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 342,00 €
    342,00 €
    1 * 1 pezzi
       342,00 €
  • 556-0027
    Complete column tube top flange with screw seal without hydraulics
    Complete column tube top flange with screw seal without hydraulics
    100,00 mm
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 32 450,00 €
    32 450,00 €
    1 * 1 pezzi
       32 450,00 €
  • 556-0010
    Complete packing stand NW100 top flange with screw seal, incl. hydraulics stand
    Complete packing stand NW100 top flange with screw seal, incl. hydraulics stand
    100,00 mm
    400 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 40 740,00 €
    40 740,00 €
    1 * 1 pezzi
       40 740,00 €
  • 556-0025
    Complete column tube top flange with screw seal without hydraulics
    Complete column tube top flange with screw seal without hydraulics
    100,00 mm
    400 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 31 480,00 €
    31 480,00 €
    1 * 1 pezzi
       31 480,00 €
  • 556-0026
    Complete column tube with temperature control jacket and top flange with screw seal without hydraulics
    Complete column tube with temperature control jacket and top flange with screw seal without hydraulics
    100,00 mm
    400 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 35 700,00 €
    35 700,00 €
    1 * 1 pezzi
       35 700,00 €
  • 556-0060
    Column tube with temperature control jacket
    Column tube with temperature control jacket
    100,00 mm
    400 mm
    Log in for availability
    Log in for availability
    1 * 1 pezzi
    1 * 1 pezzi - 8 450,00 €
    8 450,00 €
    1 * 1 pezzi
       8 450,00 €