Pullulan (from Aureobasidium pullulans) ≥96%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:9057-02-7
:
Formula:(C₆H₁₂O₅)n
MDL Number:MFCD00081940
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Presentation:White Powder
- Format:Powder
- Solubility (5% DI Water):Clear colorless solution
- Storage & Handling:Store at Room Temperature (15 to 30 °C)
- Identity Test:Passes
- Source:Aureobasidium pullulans
- Assay (dry basis):96% (of glucan)
- pH (10% soln):5,30
- Loss on drying:3,2%
- Ash:0,18%
- Viscosity (10% w/w aq. soln at 30 °C):124 mm2/s
- Lead:1 mg/kg
- Arsenic:≤0,5 mg/kg
- Total Plate Count:1000/g
- Yeast & Molds:20/g
- Coliforms:Absence/25 g
- Salmonella:Negative/25 g
Specification Test Results
Frequently Bought Together