Soy protein (from soybean), MP Biomedicals
Supplier: MP Biomedicals
Certificates
About this item
Soy protein isolated is a source of protein isolate from soybean. It has more than 92% protein content.
It is used in preparing animal research diets.
Specifications
- Source:Soybean
- Storage conditions:+15 to +30 °C
- Form:Powder
- Protein/peptide name:Soy protein
- Colour:Tan
Specifications
Frequently Bought Together