Order Entry
Italy
ContactUsLinkComponent
Blasticidin S hydrochloride (from Streptomyces griseochromogenes) biological grade
Blasticidin S hydrochloride (from Streptomyces griseochromogenes) biological grade
Catalog # A3784.0025
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Blasticidin S hydrochloride (from Streptomyces griseochromogenes) biological grade
Catalog # A3784.0025
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :458.904 g/mol
CAS Number:3513-03-9
Formula:C₁₇H₂₇ClN₈O₅
MDL Number:MFCD02091640