Order Entry
Italy
ContactUsLinkComponent
Human recombinant IFN-alpha 2b (from E. coli)
Human recombinant IFN-alpha 2b (from E. coli)
Supplier:  Genscript
Certificates
Human recombinant IFN-alpha 2b (from E. coli)
Supplier:  Genscript

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at −80 °C.
  • Reconstitution instructions:
    Reconstituted in ddH₂O at 100 μg/ml
  • Protein synonyms:
    MGC125764|Interferon alpha 2b|INFA2|MGC125765|IFNA
  • Protein/peptide name:
    IFN-alpha 2b
  • Purity:
    > 95%
  • Molecular weight:
    19,2kDa
  • Endotoxin level:
    < 0,2 EU/μg
  • Formulation:
    Lyophilized after extensive dialysis against PBS

Specifications

Frequently Bought Together