Order Entry
Italy
ContactUsLinkComponent
 
Aspergillus japonicus Pectolyase Y-23, MP Biomedicals
  ICNA08320951
 :  
Aspergillus japonicus Pectolyase Y-23, MP Biomedicals
  ICNA08320951
 :  

 

  • Pk:
    1 g
  • CAS No.:
    9033-35-6
  • Species:
    Aspergillus japonicus
  • Seq:
    01
  • Storage conditions:
    2...8 °C
  • Form:
    Light brown powder
  • Enzyme name:
    Pectolyase Y-23
  • Enzyme activity:
    >1000 units per gram
  • pH:
    5,5

 

 

Pectolyase Y-23 contains two types of pectinase such as endopolygalacturonase and endo-pectin lyase in high activity. It includes maceration stimulating factor which stimulates tissue maceration by both pectinases.

  • Soluble in water

It is used for isolation of biologically active protoplasts from a wide spectrum of higher plants and tissues in combination with a cellulase preparation in a brief incubation.

Unit definition: One unit is defined as an increase in A235 by 1,0 in the reaction mixture per minute.