Order Entry
Italy
ContactUsLinkComponent
Human EPCR ELISA kit
Human EPCR ELISA kit
Catalog # BIRBORB138076-96
Supplier:  Biorbyt
CAS Number:  
undefined
Human EPCR ELISA kit
Catalog # BIRBORB138076-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Description:
    Human EPCR ELISA Kit
  • Detection range:
    312 - 20000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

Human EPCR ELISA kit for the quantitative detection of human endothelial protein C receptor (EPCR) in cell culture supernates, serum and plasma (heparin, EDTA).

Endothelial protein C receptor (EPCR) also known as CCCA or PROCR is a protein that in humans is encoded by the PROCR gene. PRPCR gene is mapped to 20q11.2 by radiation hybrid analysis and FISH. The protein encoded by this gene is a receptor for activated protein C, a serine protease activated by and involved in the blood coagulation pathway. The encoded protein is an N-glycosylated type I membrane protein that enhances the activation of protein C. Mutations in this gene have been associated with venous thromboembolism and myocardial infarction, as well as with late fetal loss during pregnancy.