Estradiol ELISA kit
Catalog # ABNOKA4007
Supplier: Abnova
CAS Number:
Specifications
- Pk:1 KIT
- Target protein:Estriol
- Description:Estradion ELISA kit, 96 wells
Specifications
About this item
The estradiol ELISA kit is a colorimetric, solid-phase enzyme immunoassay for the quantitative detection of estradiol in plasma and serum.
- Calibration range: 0,1 nmol/l to 20 nmol/l
- Limit of detection: 0,05 nmol/l
- Sample volume: 25 µl
- Absorbance: 450 nm
Delivery: Kit contains estradiol EIA strips 96(8×12) wells, calibrator set (0; 0.1; 0.3; 1; 3, 20 nmol/l) each 0,8 ml, control serum, conjugate, substrate solution, washing solution concentrate (21×22 ml), stop solution (11 ml) and plate sealing tape (2 slides).