Order Entry
Italy
Orders LinkContactUsLinkComponent
VWR® Rota® Duo, pH indicator papers
VWR® Rota® Duo, pH indicator papers
VWR® Rota® Duo, pH indicator papers
Nome del fornitore:  VWR Chemicals
Certificates
VWR® Rota® Duo, pH indicator papers
VWR® Rota® Duo, pH indicator papers
6 Options Available Below
Product Details & Documents
Duo indicator papers show two different colours for each pH value.
  • Easy to use for qualitatively accurate readings
  • Produced from high quality paper to give good colour matching of paper to printed calibration scale
  • Narrow range papers with graduation intervals of 0.3 pH units on the colour scale
  • Wide range papers with graduation intervals up to 1 pH unit on the colour scale
Delivery information: Supplied in plactic reels that ensure protection and long-term stability. Reel of 5 m length, width: 10 mm
Product Family Options
Product Information