Protease inhibitor cocktails I, animal-free
Catalog # J65358.LQ
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 U/g
- Description:Protease inhibitor cocktail I
Specifications
About this item
A 1X stock solution contains 500 μM AEBSF.HCl, 150 nM aprotinin, 1 μM E-64, 0,5 mM EDTA and 1 μM leupeptin hemisulfate. Reconstitute each vial with 1 ml water to obtain a 100X stock solution.