Human recombinant Galectin 3 (from E. coli)
Nome del fornitore: Biorbyt
Certificati
Informazioni su questo articolo
Specifiche
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:LGALS3 although stable 4 °C for weeks, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Protein synonyms:CBP 35
- Protein/peptide name:Galectin 3
- Purity:Greater than 95.0% as determined by SDS-PAGE.
- Sequence:MGSSHHHHHH SSGLVPRGSH MADNFSLHDA LSGSGNPNPQ GWPGAWGNQP AGAGGYPGAS YPGAYPGQAP PGAYPGQAPP GAYPGAPGAY PGAPAPGVYP GPPSGPGAYP SSGQPSATGA YPATGPYGAP AGPLIVPYNL PLPGGVVPRM LITILGTVKP NANRIALDFQ RGNDVAFHFN PRFNENNRRV IVCNTKLDNN WGREERQSVF PFESGKPFKI QVLVEPDHFK VAVNDAHLLQ YNHRVKKLNE ISKLGISGDI DLTSASYTMI.
- Formulation:The Galectin-3 His tag protein solution contains 20 mM Tris-HCl pH-8, 10% glycerol 1mM DTT.
- Tested applications:SDS-PAGE , HPLC , FuncS
Specifiche
Frequently Bought Together