Mouse recombinant mRELM His (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Mouse
- Storage conditions:Store lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C.The lyophilized protein remains stable until the expiry date when stored at −20 °C.
- Protein synonyms:Resistin-like alpha
- Protein/peptide name:mRELM His
- Purity:Greater than 95% as determined by SDS-PAGE. Two-step procedure using size exclusion chromatography before and after refolding.
- Solubility:Add 0.2 uml of deionized H2O and let the lyophilized pellet dissolve completely.
- Formulation:Sterile filtered and lyophilized from 0.5 mg/ml in 5mM Tris pH 7.5, 25mM NaCl.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together