Human CD29 (from E. coli)
Catalog # BIRBORB81006-5
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:5 µG
- Source:E. coli
- Species:Human
- Storage conditions:Store vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.
- Protein synonyms:Integrin VLA-4 subunit beta
- Protein/peptide name:CD29
- Formulation:CD29 at 100ug/ml in 50mM Tris-Acetate, pH7.5, 1mM EDTA and 20% Glycerol.
- Tested applications:SDS-PAGE , HPLC , FuncS