Order Entry
Italy
ContactUsLinkComponent
 
Human recombinant TPD52L1 (from E. coli)
 :  Biorbyt
 
Human recombinant TPD52L1 (from E. coli)
 :  Biorbyt

 

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA). Avoid multiple freeze-thaw cycles.
  • Protein synonyms:
    Tumor
  • Protein/peptide name:
    TPD52L1
  • Purity:
    Greater than 90.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Sequence:
    MGSSHHHHHH SSGLVPRGSH MEAQAQGLLE TEPLQGTDED AVASADFSSM LSEEEKEELK AELVQLEDEI TTLRQVLSAK ERHLVEIKQK LGMNLMNELK QNFSKSWHDM QTTTAYKKTH ETLSHAGQKA TAAFSNVGTA ISKKFGDMRR K.
  • Formulation:
    The protein contains phosphate buffered saline, pH 7.4.
  • Tested applications:
    SDS-PAGE

 

 

Frequently Bought Together