Human Troponin 1 (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:FsTnI although stable at 10 °C for days, should be stored desiccated below −18 °C. Please prevent freeze/thaw cycles.
- Protein synonyms:Troponin fast skeletal muscle
- Protein/peptide name:Troponin 1
- Purity:Greater than 85.0% as determined by SDS-PAGE.
- Formulation:The protein solution contains 20 mM Tris-HCl, 500mM NaCl and 10mM beta mercaptoethanol, pH 7.5.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together