Order Entry
Italy
ContactUsLinkComponent
Yeast recombinant yeast TRX (from Escherichia coli)
Yeast recombinant yeast TRX (from Escherichia coli)
Supplier:  Biorbyt
Certificates
Yeast recombinant yeast TRX (from Escherichia coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Yeast
  • Storage conditions:
    TRX although stable at 4 °C for weeks, should be stored desiccated below −18 °C. Please prevent freeze thaw cycles.
  • Biological activity:
    TRX activity is assayed by measuring the change in absorbance at 650 nm at 25C using 0.13uM bovine insulin containing 0.33mM DTT (pH 6.5).The specific activity was found to be 3IU/mg.
  • Protein synonyms:
    ADF
  • Protein/peptide name:
    TRX
  • Purity:
    Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized TRX in sterile H2O.
  • Formulation:
    Each mg of protein contains 20 mM phosphate buffer pH 7.4.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together