Human HDGF (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:E. coli
- Species:Human
- Storage conditions:Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
- Protein synonyms:Hepatoma-derived growth factor
- Protein/peptide name:HDGF
- Purity:Greater than 95% as determined by SDS-PAGE.
- Sequence:MSRSNRQKEY KCGDLVFAKM KGYPHWPARI DEMPEAAVKS TANKYQVFFF GTHETAFLGP KDLFPYEESK EKFGKPNKRK GFSEGLWEIE NNPTVKASGY.
- Formulation:The HDGF protein solution is formulated in 20 mM Tris-HCl pH-7.5 1mM DTT, 10% glycerol and 1mM EDTA.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together