Order Entry
Italy
ContactUsLinkComponent
Human Recombinant 4 1BBL (from E. coli)
Human Recombinant 4 1BBL (from E. coli)
Supplier:  Biorbyt
Certificates
Human Recombinant 4 1BBL (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    4-1BBL although stable at 10 °C for weeks, should be stored desiccated below −18 °C. Please prevent freeze-thaw cycles.
  • Protein synonyms:
    TNFSF9
  • Protein/peptide name:
    4 1BBL
  • Purity:
    Greater than 95.0% as determined by SDS-PAGE
  • Sequence:
    MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMRE GPELSPDDPA GLLDLRQGMF AQLVAQNVLL IDGPLSWYSD PGLAGVSLTGGLSYKEDTKE LVVAKAGVYY VFFQLELRRV VAGEGSGSVS LALHLQPLRS AAGAAALALT VDLPPASSEA RNSAFGFQGR LLHLSAGQRL GVHLHTEARA RHAWQLTQGA TVLGLFRVTP EIPAGLPSPR SE.
  • Formulation:
    Recombinant 4-1BBL contains 20 mM Tris buffer pH-8, 100mM NaCl and 20% glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together