Human recombinant CNTF His (from E. coli)
: Biorbyt
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:CNTF his tag fusion protein although stable 4 °C for week, should be stored desiccated below −18 °C. Please prevent freeze-thaw cycles.
- Protein synonyms:HCNTF
- Protein/peptide name:CNTF His
- Purity:Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Sequence:MGSSHHHHHH SSGLVPRGSH MAFTEHSPLT PHRRDLCSRS IWLARKIRSDLTALTESYVK HQGLNKNINL DSADGMPVAS TDQWSELTEA ERLQENLQAY RTFHVLLARL LEDQQVHFTP TEGDFHQAIH TLLLQVAAFA YQIEELMILL EYKIPRNEAD GMPINVGDGG LFEKKLWGLK VLQELSQWTV RSIHDLRFIS SHQTGIPARG SHYIANNKKM.
- Formulation:CNTF His tag protein solution contains 20 mM Tris buffer pH-8 and 1mM DTT.
- Tested applications:SDS-PAGE , HPLC , FuncS
Frequently Bought Together