Order Entry
Italy
ContactUsLinkComponent
 
Human recombinant BAFF (from E. coli)
 :  Biorbyt
 
Human recombinant BAFF (from E. coli)
 :  Biorbyt

 

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    BLyS although stable at 10 °C for weeks, should be stored desiccated below −18 °C. Please prevent freeze-thaw cycles.
  • Protein/peptide name:
    BAFF
  • Purity:
    Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Sequence:
    MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMAV QGPEETVTQDCLQLIADSET PTIQKGSYTF VPWLLSFKRG SALEEKENKI LVKETGYFFI YGQVLYTDKT YAMGHLIQRK KVHVFGDELS LVTLFRCIQN MPETLPNNSC YSAGIAKLEE GDELQLAIPR ENAQISLDGD VTFFGALKLL.
  • Formulation:
    Recombinant His Tag BAFF contains 20 mM Tris buffer pH-8 and 5mM DTT.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

 

 

Frequently Bought Together