Human recombinant Pleiotrophin (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C.The lyophilized protein remains stable until the expiry date when stored at −20 °C.
- Protein synonyms:PTN
- Protein/peptide name:Pleiotrophin
- Purity:Greater than 95% as determined by SDS-PAGE. Three-step procedure using affinity Ni-NTA chromatography and size exclusion chromatography before and after refolding.
- Solubility:Add 0.2 uml of PBS pH 7.2 and let the lyophilized pellet dissolve completely.
- Formulation:Sterile filtered and lyophilized from 0.5 mg/ml in 0.1M phosphate buffer and 0.1M NaCl, pH 7.2.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together