Human recombinant CD99 (from E. coli)
: Biorbyt
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store vial at −20 °C to −80 °C. When stored at the recommended temperature, this protein is stable for One year.Please prevent freeze-thaw cycles.
- Protein synonyms:CD99 antigen
- Protein/peptide name:CD99
- Formulation:CD99 at 0.1mg/ml in 50mM Tris-Acetate, pH7.5, 1mM EDTA and 20% Glycerol.
- Tested applications:SDS-PAGE , HPLC , FuncS
Frequently Bought Together