Order Entry
Italy
ContactUsLinkComponent
Mouse recombinant cystatin C (from E. coli)
Mouse recombinant cystatin C (from E. coli)
Supplier:  Biorbyt
Certificates
Mouse recombinant cystatin C (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Mouse
  • Storage conditions:
    Store lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time.
  • Protein synonyms:
    AD8
  • Protein/peptide name:
    Cystatin C
  • Purity:
    Greater than 95% as determined by SDS-PAGE.
  • Solubility:
    Add 0.1M Acetate buffer pH to working concentration of 0.5mg/ml and let the lyophilized pellet dissolve completely.
  • Sequence:
    MRGSHHHHHH GMASATPKQG PRMLGAPEEA DANEEGVRRA LDFAVSEYNK GSNDAYHSRA IQVVRARKQL VAGVNYFLDV EMGRTTCTKS QTNLTDCPFH DQPHLMRKAL CSFQIYSVPW KGTHSLTKFSCKNA.
  • Formulation:
    The sterile filtered concentrated (0.5mg/ml) protein solution was lyophilized with 20 mM Tris 50mM NaCl pH-7.5.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together