Human recombinant CD62P (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
- Protein/peptide name:CD62P
- Purity:Greater than 95% as determined by SDS-PAGE. Single band on Western Blot.
- Formulation:P-Selectin is supplied in 50% Glycerol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together