Order Entry
Italy
ContactUsLinkComponent
 
Human VEGF receptor 1 D1-3 (from Insect cells)
 :  Biorbyt
 
Human VEGF receptor 1 D1-3 (from Insect cells)
 :  Biorbyt

 

  • Source:
    Insect cells
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. Please avoid freeze thaw cycles.
  • Protein synonyms:
    Vascular endothelial growth factor A
  • Protein/peptide name:
    VEGF receptor 1 D1-3
  • Purity:
    Greater than 95.0% as determined by(a)Analysis by RP-HPLC. (b)Analysis by SDS-PAGE Usage Biorbyt's products are furnished for LABORATORY RESEARCH USE ONLY. The product may not be used as drugs, agricultural or pesticidal products, food additives or househ
  • Formulation:
    FLT1 His is supplied in 1x PBS and 50% glycerol.
  • Tested applications:
    SDS-PAGE

 

 

Frequently Bought Together