Mouse TIE-1 Fc chimera (from CHO cells)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:CHO cells
- Species:Mouse
- Storage conditions:Lyophilized sTIE-1 although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution TIE-1 should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Protein synonyms:Tyrosine kinase with immunoglobulin-like and EGF-like domains 1
- Protein/peptide name:TIE-1 Fc Chimera
- Purity:Greater than 90.0% as determined by(a)Analysis by RP-HPLC.(b)Analysis by SDS-PAGE.
- Solubility:It is recommended to reconstitute the lyophilized TIE-1 Fc Chimera in sterile water not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Formulation:TIE-1 Fc Chimera was lyophilized from concentrated (1 mg/ml) sterile solution containing 1x PBS.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together