HIV-1 p66 pol (from Baculovirus (insect cells))
: Biorbyt
- Source:Baculovirus (Insect cells)
- Storage conditions:Recombinant HIV-1 p66 although stable at 4 °C for weeks, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA). Please avoid freeze-thaw cycles.
- Protein/peptide name:HIV-1 p66 pol
- Purity:Greater than 90.0% as determined by HPLC analysis SDS-PAGE.
- Formulation:The HIV-1 p66 pol protein solution contains 30mM Tris pH-7, 0.15M NaCl, 0.2 um EDTA 2mM b-ME.
- Tested applications:SDS-PAGE
Frequently Bought Together