Order Entry
Italy
ContactUsLinkComponent
Human recombinant APOM (from E. coli)
Human recombinant APOM (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant APOM (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    APOM Human although stable at 4 °C for week, should be stored desiccated below −18 °C. Please prevent freeze thaw cycles.
  • Protein synonyms:
    G3a
  • Protein/peptide name:
    APOM
  • Purity:
    Greater than 90% as determined by SDS-PAGE.
  • Sequence:
    MGSSHHHHHH SSGLVPRGSH MCPEHSQLTT LGVDGKEFPE VHLGQWYFIA GAAPTKEELA TFDPVDNIVF NMAAGSAPMQ LHLRATIRMK DGLCVPRKWI YHLTEGSTDL RTEGRPDMKT ELFSSSCPGG IMLNETGQGY QRFLLYNRSP HPPEKCVEEF KSLTSCLDSK AFLLTPRNQE ACELSNN.
  • Formulation:
    APOM Human solution containing 20 mM Trsi HCL pH-8 1mM DTT 10% glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together