Order Entry
Italy
ContactUsLinkComponent
Human recombinant Osteopontin (from E. coli)
Human recombinant Osteopontin (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant Osteopontin (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time. Please avoid freeze thaw cycles.
  • Protein synonyms:
    BNSP
  • Protein/peptide name:
    Osteopontin
  • Purity:
    Greater than 90.0% as determined by SDS-PAGE.
  • Sequence:
    MGSSHHHHHH SSGLVPRGSH RSMIPVKQAD SGSSEEKQLY NKYPDAVATW LNPDPSQKQN LLAPQNAVSS EETNDFKQET LPSKSNESHD HMDDMDDEDD DDHVDSQDSI DSNDSDDVDD TDDSHQSDES HHSDESDELV TDFPTDLPAT EVFTPVVPTV DTYDGRGDSV VYGLRSKSKK FRRPDIQYPD ATDEDITSHM ESEELNGAYK AIPVAQDLNA PSDWDSRGKD SYETSQLDDQ SAETHSHKQS RLYKRKANDE SNEHSDVIDS QELSKVSREF HSHEFHSHED MLVVDPKSKE EDKHLKFRIS HELDSASSEV N.
  • Formulation:
    Osteopontin is supplied in 20 mM Tris-HCL buffer pH-7.5, 1mM DTT, 2mM EDTA and 10% glycerol.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together