Order Entry
Italy
ContactUsLinkComponent
Human recombinant RBP4 His (from E. coli)
Human recombinant RBP4 His (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant RBP4 His (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Lyophilized Retinol binding protein-4 although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution Retinol binding protein-4 should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Biological activity:
    Determined by its ability to bind all trans retinol acids. The binding of RBP4 to all trans retinol acids results in quenching of Trp fluroscence. 0.5M retinol acids/per RBP4.
  • Protein synonyms:
    Retinol Binding
  • Protein/peptide name:
    RBP4 His
  • Purity:
    Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Retinol binding protein-4 in sterile deionized water to working volume of 0.5mg/mL and let the lyophilized pellet dissolve completely. Product is not sterile! Please filter the product by an appropriate st
  • Sequence:
    MSWWHHHHHH NWNIPTTQDT TERDCRVSSF RVKENFDKAR FSGTWYAMAK KDPEGLFLQD NIVAEFSVDE TGQMSATAKG RVRLLNNWDV CADMVGTFTD TEDPAKFKMK YWGVASFLQK GNDDHWIVDT DYDTYAVQYS CRLLNLDGTC ADSYSFVFSR DPNGLPPEAQ KIVRQRQEEL CLARQYRLIV HNGYCDGRSERNLL.
  • Formulation:
    RBP-4 His tag was filtered (0.4 um) and lyophilized from 0.5 mg/ml in PBS buffer, pH 7.5.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together