Mouse adiponectin glycosilated HEK (from HEK293 cells)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:HEK293 cells
- Species:Mouse
- Storage conditions:Store lyophilized APM-1 at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C.The lyophilized protein remains stable until the expiry date when stored at −20 °C.
- Biological activity:Full-length mouse adiponectin activates AMP-activated protein kinase in hepatocyte and activates AMPK in HepG2 human hepatocytes at concentration of 1ug/ml.Mouse adiponectin mammalian cell derived inhibits glucose production as shown by in-vitro gluconeogenesis assay in primary rat hepatocytes.
- Gene ID:11450
- Protein synonyms:ACDC
- UniProtKB:Q60994 (Mouse)
- Protein/peptide name:Adiponectin glycosilated HEK
- Purity:Purity of adiponectin mouse recombinant is >98% (HPLC and SDS PAGE analyzed).
- Solubility:Add 0.2 uml of deionized water and let the lyophilized pellet dissolve completely.
- Sequence:EDDVTTTEEL APALVPPPKG TCAGWMAGIP GHPGHNGTPG RDGRDGTPGE KGEKGDAGLL GPKGETGDVG MTGAEGPRGF PGTPGRKGEP GEAAYMYRSA FSVGLETRVT VPNVPIRFTK IFYNQQNHYD GSTGKFYCNI PGLYYFSYHI TVYMKDVKVS LFKKDKAVLF TYDQYQEKNV DQASGSVLLH LEVGDQVWLQ VYGDGDHNGL YADNVNDSTF TGFLLYHDTN DYKDDDDK.
- Formulation:Sterile filtered and lyophilized from 0.5 mg/ml in PBS buffer.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together