VEGF-E (Orf virus) (from Escherichia coli)
: Biorbyt
- Source:E. coli
- Storage conditions:Lyophilized Vascular Endothelial Growth Factor-E Orf Virus although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution VEGF -OV should be stored at 4 °C between 2-7 days and for future use below −18 °C. Please prevent freeze/thaw cycles.
- Biological activity:The ED50 for stimulation of 3H-thymidine incorporation and cell proliferation by human umbilical vein endothelial cells for VEGF-E Orf Virus has been determined to be in the range of - 20 ng/ml, corresponding to specific activity of 200,000 Units/mg.
- Protein/peptide name:VEGF-E (Orf Virus)
- Purity:Greater than 90.0% as determined by RP-HPLC and analysis by SDS-PAGE
- Solubility:The lyophilized VEGF-E is soluble in water and most aqueous buffers. The lyophilized Vascular Endothelial Growth Factor-E (Orf Virus) should be reconstituted in PBS or medium containing at least 0.1% human or bovine serum albumin to concentration not lowe
- Formulation:The protein was lyophilized from concentrated (1mg/ml) solution containing PBS pH-7.4.
- Tested applications:SDS-PAGE
Frequently Bought Together