Order Entry
Italy
ContactUsLinkComponent
Human recombinant IL6 (from Insect cells)
Human recombinant IL6 (from Insect cells)
Supplier:  Biorbyt
Certificates
Human recombinant IL6 (from Insect cells)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    Insect cells
  • Species:
    Human
  • Storage conditions:
    Lyophilized Hepatocyte Growth Factor although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution HGF should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
  • Biological activity:
    The activity was assayed for scattering activity in the MDCK cell assay.The ED50 for this effect is typically at 0.5-1.0 ng/ml.
  • Protein synonyms:
    Interleukin BSF 2
  • Protein/peptide name:
    IL6
  • Purity:
    Greater than 90.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Solubility:
    It is recommended to reconstitute the lyophilized Hepatocyte Growth Factor in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Sequence:
    Agrees with the sequence of native human HGF.
  • Formulation:
    The sterile protein powder (1 mg/ml) is lyophilized from solution containing 50mM Acetic Acid.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together